Merck
  • A Common Embryonic Origin of Stem Cells Drives Developmental and Adult Neurogenesis.
A Common Embryonic Origin of Stem Cells Drives Developmental and Adult Neurogenesis.
Cell (2019-04-02)
Daniel A Berg, Yijing Su, Dennisse Jimenez-Cyrus, Aneek Patel, Nancy Huang, David Morizet, Stephanie Lee, Reeti Shah, Francisca Rojas Ringeling, Rajan Jain, Jonathan A Epstein, Qing-Feng Wu, Stefan Canzar, Guo-Li Ming, Hongjun Song, Allison M Bond
ABSTRACT
New neurons arise from quiescent adult neural progenitors throughout life in specific regions of the mammalian brain. Little is known about the embryonic origin and establishment of adult neural progenitors. Here, we show that Hopx+ precursors in the mouse dentate neuroepithelium at embryonic day 11.5 give rise to proliferative Hopx+ neural progenitors in the primitive dentate region, and they, in turn, generate granule neurons, but not other neurons, throughout development and then transition into Hopx+ quiescent radial glial-like neural progenitors during an early postnatal period. RNA-seq and ATAC-seq analyses of Hopx+ embryonic, early postnatal, and adult dentate neural progenitors further reveal common molecular and epigenetic signatures and developmental dynamics. Together, our findings support a "continuous" model wherein a common neural progenitor population exclusively contributes to dentate neurogenesis throughout development and adulthood. Adult dentate neurogenesis may therefore represent a lifelong extension of development that maintains heightened plasticity in the mammalian hippocampus.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-Prox1 Antibody, clone 4G10, clone 4G10, Chemicon®, from mouse