Merck
  • GPD1 Specifically Marks Dormant Glioma Stem Cells with a Distinct Metabolic Profile.
GPD1 Specifically Marks Dormant Glioma Stem Cells with a Distinct Metabolic Profile.
Cell stem cell (2019-07-16)
Patricia Rusu, Chunxuan Shao, Anna Neuerburg, Azer Aylin Acikgöz, Yonghe Wu, Peng Zou, Prasad Phapale, Tchirupura S Shankar, Kristina Döring, Steffen Dettling, Huiqin Körkel-Qu, Gözde Bekki, Barbara Costa, Te Guo, Olga Friesen, Magdalena Schlotter, Mathias Heikenwalder, Darjus F Tschaharganeh, Bernd Bukau, Günter Kramer, Peter Angel, Christel Herold-Mende, Bernhard Radlwimmer, Hai-Kun Liu
ABSTRACT
Brain tumor stem cells (BTSCs) are a chemoresistant population that can drive tumor growth and relapse, but the lack of BTSC-specific markers prevents selective targeting that spares resident stem cells. Through a ribosome-profiling analysis of mouse neural stem cells (NSCs) and BTSCs, we find glycerol-3-phosphate dehydrogenase 1 (GPD1) expression specifically in BTSCs and not in NSCs. GPD1 expression is present in the dormant BTSC population, which is enriched at tumor borders and drives tumor relapse after chemotherapy. GPD1 inhibition prolongs survival in mouse models of glioblastoma in part through altering cellular metabolism and protein translation, compromising BTSC maintenance. Metabolomic and lipidomic analyses confirm that GPD1+ BTSCs have a profile distinct from that of NSCs, which is dependent on GPD1 expression. Similar GPD1 expression patterns and prognostic associations are observed in human gliomas. This study provides an attractive therapeutic target for treating brain tumors and new insights into mechanisms regulating BTSC dormancy.
Sigma-Aldrich
Anti-PCNA (Ab-1) Mouse mAb (PC10), liquid, clone PC10, Calbiochem®
Sigma-Aldrich
Anti-ANXA6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CNPase Antibody, clone 11-5B, clone 11-5B, Chemicon®, from mouse
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse