Merck
  • LncRNA-NEF is downregulated in postmenopausal osteoporosis and is related to course of treatment and recurrence.
LncRNA-NEF is downregulated in postmenopausal osteoporosis and is related to course of treatment and recurrence.
The Journal of international medical research (2019-06-22)
Xiaoyong Ma, Zhixue Guo, Wenshan Gao, Jianzhong Wang, Yong Liu, Fei Gao, Shaosong Sun, Xiaozhe Zhou, Zhaoyu Yang, Wenkui Zheng
Sigma-Aldrich
Human Mesenchymal Stem Cells (Bone Marrow), Human mesenchymal stem cells derived from bone marrow that have a capacity for expansion in vitro while maintaining their potential for differentiation to multiple lineages including adipocytes, osteoblasts and chondrocytes.