Merck
  • Fused Heterocyclic Systems with an s-Triazine Ring. 34. Development of a Practical Approach for the Synthesis of 5-Aza-isoguanines.
Fused Heterocyclic Systems with an s-Triazine Ring. 34. Development of a Practical Approach for the Synthesis of 5-Aza-isoguanines.
Molecules (Basel, Switzerland) (2019-04-25)
Ahmad Junaid, Felicia Phei Lin Lim, Yvonne Peijun Zhou, Wai Keung Chui, Anton V Dolzhenko
ABSTRACT
Purine isosteres present excellent opportunities in drug design and development. Using isosteres of natural purines as scaffolds for the construction of new therapeutic agents has been a valid strategy of medicinal chemistry. Inspired by the similarity to isoguanine, we attempted to develop a practical method for the preparation of 5-aza-isoguanines. Several synthetic approaches were explored to establish a robust general protocol for the preparation of these compounds. The significant difference in the reactivity of the C-5 and C-7 electrophilic centers of 1,2,4-triazolo[1,5-a][1,3,5]triazines (5-azapurines) towards nucleophiles was demonstrated. The most practical and general method for the preparation of 5-aza-isoguanines involved a regioselective reaction of ethoxycarbonyl isothiocyanate with a 5-aminotriazole. The intramolecular ring closure of the resulted product followed by the S-methylation afforded 7-methylthio-2-phenyl-1,2,4-triazolo[1,5-a][1,3,5]triazin-5-one, which could be effectively aminated with various amines. The resulted 5-aza-isoguanines resemble a known purine nucleoside phosphorylase inhibitor and could be interesting for further investigations as potential anticancer agents.