Merck
Weak magnetic fields alter stem cell-mediated growth.
Science advances (2019-02-08)
Alanna V Van Huizen, Jacob M Morton, Luke J Kinsey, Donald G Von Kannon, Marwa A Saad, Taylor R Birkholz, Jordan M Czajka, Julian Cyrus, Frank S Barnes, Wendy S Beane
ABSTRACT
Biological systems are constantly exposed to electromagnetic fields (EMFs) in the form of natural geomagnetic fields and EMFs emitted from technology. While strong magnetic fields are known to change chemical reaction rates and free radical concentrations, the debate remains about whether static weak magnetic fields (WMFs; <1 mT) also produce biological effects. Using the planarian regeneration model, we show that WMFs altered stem cell proliferation and subsequent differentiation via changes in reactive oxygen species (ROS) accumulation and downstream heat shock protein 70 (Hsp70) expression. These data reveal that on the basis of field strength, WMF exposure can increase or decrease new tissue formation in vivo, suggesting WMFs as a potential therapeutic tool to manipulate mitotic activity.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, clone MC463, rabbit monoclonal, culture supernatant, clone MC463, Upstate®
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%