Merck
  • Long-term tolerance of islet allografts in nonhuman primates induced by apoptotic donor leukocytes.
Long-term tolerance of islet allografts in nonhuman primates induced by apoptotic donor leukocytes.
Nature communications (2019-08-04)
Amar Singh, Sabarinathan Ramachandran, Melanie L Graham, Saeed Daneshmandi, David Heller, Wilma Lucia Suarez-Pinzon, Appakalai N Balamurugan, Jeffrey D Ansite, Joshua J Wilhelm, Amy Yang, Ying Zhang, Nagendra P Palani, Juan E Abrahante, Christopher Burlak, Stephen D Miller, Xunrong Luo, Bernhard J Hering
ABSTRACT
Immune tolerance to allografts has been pursued for decades as an important goal in transplantation. Administration of apoptotic donor splenocytes effectively induces antigen-specific tolerance to allografts in murine studies. Here we show that two peritransplant infusions of apoptotic donor leukocytes under short-term immunotherapy with antagonistic anti-CD40 antibody 2C10R4, rapamycin, soluble tumor necrosis factor receptor and anti-interleukin 6 receptor antibody induce long-term (≥1 year) tolerance to islet allografts in 5 of 5 nonsensitized, MHC class I-disparate, and one MHC class II DRB allele-matched rhesus macaques. Tolerance in our preclinical model is associated with a regulatory network, involving antigen-specific Tr1 cells exhibiting a distinct transcriptome and indirect specificity for matched MHC class II and mismatched class I peptides. Apoptotic donor leukocyte infusions warrant continued investigation as a cellular, nonchimeric and translatable method for inducing antigen-specific tolerance in transplantation.