Merck
  • Anti-biofilm and cytoprotective activities of quercetin against Pseudomonas aeruginosa isolates.
Anti-biofilm and cytoprotective activities of quercetin against Pseudomonas aeruginosa isolates.
Letters in applied microbiology (2019-02-15)
C Vipin, M Mujeeburahiman, P Ashwini, A B Arun, P-D Rekha
ABSTRACT
Increase in infection with multidrug resistant Pseudomonas aeruginosa is a serious global challenge in healthcare. Pseudomonas aeruginosa is capable of causing human infection in various sites and complicates the infection due to its virulence factors. This study was aimed to investigate the effect of quercetin, a dietary flavonoid against the virulence factors of P. aeruginosa and its cell protective effects on epithelial cells. Bactericidal activity, anti-biofilm activity and effect on different virulence factors were carried out using standard methods by using five P. aeruginosa isolates. Cytotoxicity and cell protective effect of quercetin was evaluated by trypan blue dye exclusion assay. All the tested isolates were completely inhibited (100%) by quercetin at a concentration of 500 μg ml-1 . It showed significant (P < 0·05) inhibitory effect on virulence factors including biofilm formation and showed significant protective effect on HEK 293T cells infected with P. aeruginosa strains. This study supports the role of quercetin against P. aeruginosa, by inhibiting virulence factors as well as its cytoprotective activity during bacterial infection either by attenuating the virulence or providing direct protective effect to the host cells. SIGNIFICANCE AND IMPACT OF THE STUDY: The increase in infections caused by opportunistic pathogen Pseudomonas aeruginosa is a serious concern in the health care system. This study describes the beneficial effects of a dietary flavonoid, quercetin against pathogenic P. aeruginosa strains and its protective effect against the P. aeruginosa infection in HEK 293T cells in vitro.