Merck
  • Liposomal paclitaxel induces fewer hematopoietic and cardiovascular complications than bioequivalent doses of Taxol.
Liposomal paclitaxel induces fewer hematopoietic and cardiovascular complications than bioequivalent doses of Taxol.
International journal of oncology (2018-06-30)
Shih-Ting Huang, Yi-Ping Wang, Yen-Hui Chen, Chin-Tarng Lin, Wen-Shan Li, Han-Chung Wu
ABSTRACT
Paclitaxel (PTX) exhibits potent antineoplastic activity against various human malignancies; however, clinical application must overcome the inherent hydrophobicity of this molecule. The commercialized Taxol formulation utilizes Cremophor EL (CrEL)/ethanol as a solvent to stabilize and dispense PTX in an aqueous solution. However, adverse CrEL‑induced hypersensitivity reactions have been reported in ~30% of recipients, and 40% of patients receiving premedication may also experience this adverse effect. Therefore, the development of a CrEL-free delivery system is crucial, in order to fully exploit the therapeutic efficacy of PTX. In the present study, a novel liposomal PTX (lipo‑PTX) formulation was optimized with regards to encapsulation rate and long‑term stability, arriving at a molar constituent ratio of soybean phosphatidylcholine:cholesterol:N-(carbonyl-methoxy-poly-ethylene glycol 2000)‑1,2‑distearoyl‑sn-glycero‑3-phosphoethanolamine, sodium salt:PTX at 95:2:1:2. Comparable doses of lipo‑PTX and Taxol were bioequivalent in terms of therapeutic efficacy in xenograft tumor models. However, the systemic side effects, including hematopoietic toxicity, acute hypersensitivity reactions and cardiac irregularities, were significantly reduced in lipo‑PTX‑treated mice compared with those infused with reference formulations of PTX. In conclusion, the present study reported that lipo‑PTX exhibited a higher therapeutic index than clinical PTX formulations.
Product Description
Avanti
16:0 Liss Rhod PE, Avanti Research - A Croda Brand
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Hematoxylin Solution, Gill No. 1
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)