Merck
  • Selectins: interpreters of cell-specific carbohydrate information during inflammation.
Selectins: interpreters of cell-specific carbohydrate information during inflammation.
Science (New York, N.Y.) (1992-11-06)
L A Lasky
ABSTRACT
Although a bewildering array of cell surface carbohydrate structures have been described, the physiological relevance of any of these complex molecules has often eluded biologists. A family of cell surface glycoproteins, the "selectins," has a characteristic ability to use some of these carbohydrate structures in adhesive mechanisms that help localize leukocytes to regions of inflammation. This article will review the biology of these carbohydrate-binding adhesive proteins and discuss the potential for developing anti-inflammatory antagonists that could inhibit binding events that are selectin-mediated.