Merck
  • Aprotinin interacts with substrate-binding site of human dipeptidyl peptidase III
Aprotinin interacts with substrate-binding site of human dipeptidyl peptidase III
Journal of Biomolecular Structure & Dynamics (2019)
Agic D, et al.