Merck
  • Enzymatic synthesis of nucleosides by nucleoside phosphorylase co-expressed in Escherichia coli
Enzymatic synthesis of nucleosides by nucleoside phosphorylase co-expressed in Escherichia coli
Journal of Zhejiang University. Science. B (2010)
Ding QB, et al.