Merck
  • In vitro antiproliferative activity of fresh pineapple juices on ovarian and colon cancer cell lines
In vitro antiproliferative activity of fresh pineapple juices on ovarian and colon cancer cell lines
International Journal of Peptide Research and Therapeutics (2015)
Gani MBA, et al.