- A UV spectrophotometric method for the determination of folic acid in pharmaceutical tablets and dissolution tests
A UV spectrophotometric method for the determination of folic acid in pharmaceutical tablets and dissolution tests
Analytical Methods : Advancing Methods and Applications (2014)
Matias R, et al.