Merck
Zwitterionic Acetylated Cellulose Nanofibrils.
Molecules (Basel, Switzerland) (2019-09-01)
Jowan Rostami, Aji P Mathew, Ulrica Edlund
ABSTRACT
A strategy is devised to synthesize zwitterionic acetylated cellulose nanofibrils (CNF). The strategy included acetylation, periodate oxidation, Schiff base reaction, borohydride reduction, and a quaternary ammonium reaction. Acetylation was performed in glacial acetic acid with a short reaction time of 90 min, yielding, on average, mono-acetylated CNF with hydroxyl groups available for further modification. The products from each step were characterized by FTIR spectroscopy, ζ-potential, SEM-EDS, AFM, and titration to track and verify the structural changes along the sequential modification route.
Sigma-Aldrich
3-(Dimethylamino)-1-propylamine, 99%
Sigma-Aldrich
Sodium 2-bromoethanesulfonate, 98%