Merck
  • The impact of backbone N-methylation on the structure-activity relationship of Leu10 -teixobactin.
The impact of backbone N-methylation on the structure-activity relationship of Leu10 -teixobactin.
Journal of peptide science : an official publication of the European Peptide Society (2019-08-08)
Tony Velkov, James D Swarbrick, Maytham H Hussein, Elena K Schneider-Futschik, Daniel Hoyer, Jian Li, John A Karas
ABSTRACT
Antimicrobial resistance is a serious threat to global human health; therefore, new anti-infective therapeutics are required. The cyclic depsi-peptide teixobactin exhibits potent antimicrobial activity against several Gram-positive pathogens. To study the natural product's mechanism of action and improve its pharmacological properties, efficient chemical methods for preparing teixobactin analogues are required to expedite structure-activity relationship studies. Described herein is a synthetic route that enables rapid access to analogues. Furthermore, our new N-methylated analogues highlight that hydrogen bonding along the N-terminal tail is likely to be important for antimicrobial activity.
Sigma-Aldrich
Methyl 4-nitrobenzenesulfonate, 99%
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
2,2′-(Ethylenedioxy)diethanethiol, 95%