Merck
  • Adipokine zinc-α2-glycoprotein alleviates lipopolysaccharide-induced inflammatory responses through the β3-AR/PKA/CREB pathway.
Adipokine zinc-α2-glycoprotein alleviates lipopolysaccharide-induced inflammatory responses through the β3-AR/PKA/CREB pathway.
Cytokine (2019-07-02)
Jun Guo, Yanfei Li, Ruqian Zhao, Xiaojing Yang
ABSTRACT
Humans and animals frequently experience dysmetabolism induced by inflammation. Zinc-α2-glycoprotein (ZAG), a newly identified adipokine, is potentially involved in lipid metabolism. Our previous study revealed that the ZAG content increased after lipopolysaccharide (LPS) treatment. To clarify ZAG's possible effects on inflammatory responses and lipid metabolism, we used gene overexpression and knockout mice as models to investigate the function of ZAG during inflammation. The results showed that LPS increased plasma triglyceride, non-esterified fatty acid and hepatic triglyceride, while ZAG overexpression decreased these effects. Furthermore, ZAG overexpression weakened inflammatory responses, suppressed lipogenesis, and improved mitochondrial function during inflammation. ZAG overexpression also increased β3-adrenoreceptor, protein kinase A, and phosphorylated cyclic adenosine monophosphate-response element binding protein (CREB), promoted the combination of CREB and CREB-binding protein (CBP), and competitively inhibited the combination of nuclear factor-κB and CBP. After ZAG knockout, LPS-induced the hyperlipidemia worsened. ZAG knockout aggravated inflammatory responses, promoted lipogenesis, and weakened mitochondrial function during inflammation. ZAG knockout also decreased β3-adrenoreceptor and protein kinase A. The present study demonstrated that ZAG alleviated lipid metabolism disorders by weakening inflammatory responses. The β3-adrenoreceptor/protein kinase A/CREB pathway mediated the effects of ZAG on inflammation. These results will provide new insight for research on anti-inflammation.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction