Merck
  • Interaction of α Carboxyl Terminus 1 Peptide With the Connexin 43 Carboxyl Terminus Preserves Left Ventricular Function After Ischemia-Reperfusion Injury.
Interaction of α Carboxyl Terminus 1 Peptide With the Connexin 43 Carboxyl Terminus Preserves Left Ventricular Function After Ischemia-Reperfusion Injury.
Journal of the American Heart Association (2019-08-20)
Jingbo Jiang, Daniel Hoagland, Joseph A Palatinus, Huamei He, Jegan Iyyathurai, L Jane Jourdan, Geert Bultynck, Zhen Wang, Zhiwei Zhang, Kevin Schey, Steven Poelzing, Francis X McGowan, Robert G Gourdie
ABSTRACT
Background α Carboxyl terminus 1 (αCT1) is a 25-amino acid therapeutic peptide incorporating the zonula occludens-1 (ZO-1)-binding domain of connexin 43 (Cx43) that is currently in phase 3 clinical testing on chronic wounds. In mice, we reported that αCT1 reduced arrhythmias after cardiac injury, accompanied by increases in protein kinase Cε phosphorylation of Cx43 at serine 368. Herein, we characterize detailed molecular mode of action of αCT1 in mitigating cardiac ischemia-reperfusion injury. Methods and Results To study αCT1-mediated increases in phosphorylation of Cx43 at serine 368, we undertook mass spectrometry of protein kinase Cε phosphorylation assay reactants. This indicated potential interaction between negatively charged residues in the αCT1 Asp-Asp-Leu-Glu-Iso sequence and lysines (Lys345, Lys346) in an α-helical sequence (helix 2) within the Cx43-CT. In silico modeling provided further support for this interaction, indicating that αCT1 may interact with both Cx43 and ZO-1. Using surface plasmon resonance, thermal shift, and phosphorylation assays, we characterized a series of αCT1 variants, identifying peptides that interacted with either ZO-1-postsynaptic density-95/disks large/zonula occludens-1 2 or Cx43-CT, but with limited or no ability to bind both molecules. Only peptides competent to interact with Cx43-CT, but not ZO-1-postsynaptic density-95/disks large/zonula occludens-1 2 alone, prompted increased pS368 phosphorylation. Moreover, in an ex vivo mouse model of ischemia-reperfusion injury, preischemic infusion only with those peptides competent to bind Cx43 preserved ventricular function after ischemia-reperfusion. Interestingly, a short 9-amino acid variant of αCT1 (αCT11) demonstrated potent cardioprotective effects when infused either before or after ischemic injury. Conclusions Interaction of αCT1 with the Cx43, but not ZO-1, is correlated with cardioprotection. Pharmacophores targeting Cx43-CT could provide a translational approach to preserving heart function after ischemic injury.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Anti-Connexin-43 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Millipore
ReBlot Plus Strong Antibody Stripping Solution, 10x
Sigma-Aldrich
Brilliant Blue G, pure
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, microbial, BioReagent, suitable for cell culture, ≥99% (HPLC)