Merck
  • Surfactant-Sensitized Covalent Organic Frameworks-Functionalized Lanthanide-Doped Nanocrystals: An Ultrasensitive Sensing Platform for Perfluorooctane Sulfonate.
Surfactant-Sensitized Covalent Organic Frameworks-Functionalized Lanthanide-Doped Nanocrystals: An Ultrasensitive Sensing Platform for Perfluorooctane Sulfonate.
ACS omega (2019-10-09)
Jing Li, Caiyun Zhang, Mingyuan Yin, Zhen Zhang, Yujie Chen, Qiliang Deng, Shuo Wang
ABSTRACT
Perfluorooctane sulfonate (PFOS) known as a persistent organic pollutant has been attracting great interests due to its potential ecotoxicity. An approach capable of sensing ultra-trace PFOS is in urgent demand. Here, we developed an approach for highly sensitive sensing PFOS using surfactant-sensitized covalent organic frameworks (COFs)-functionalized upconversion nanoparticles (UCNPs) as a fluorescent probe. COFs-functionalized UCNPs (UCNPs@COFs) were obtained by solvothermal growth of 1,3,5-triformylbenzene and 1,4-phenylenediamine on the surface of UCNPs. COF's layer on the surface of UCNPs not only provides recognition sites for PFOS but also improves the fluorescence quantum yields from 2.15 to 5.12%. Trace PFOS can quench the fluorescence emission of UCNPs@COFs at 550 nm due to the high electronegativity of PFOS. Moreover, the fluorescence quenching response can be significantly strengthened in the presence of a surfactant, which causes more sensitivity. The fluorescence quenching degrees (F0 - F) of the system are linear with the concentration of PFOS in the range of 1.8 × 10-13 to 1.8 × 10-8 M. The present sensor can sensitively and selectively detect PFOS in tap water and food packing with the limit of detection down to 0.15 pM (signal-to-noise ratio = 3), which is comparable to that of the liquid chromatography-mass spectrometry technique. The proposed approach realized a simple, fast, sensitive, and selective sensing PFOS, showing potential applications in various fields.
Sigma-Aldrich
Benzene-1,3,5-tricarboxaldehyde, 97%
Sigma-Aldrich
Perfluoroheptanoic acid, ≥97.0%
Sigma-Aldrich
Perfluorononanoic acid, 97%
Sigma-Aldrich
Perfluorodecanoic acid, 97%