Merck
  • Inside out: optimization of lipid nanoparticle formulations for exterior complexation and in vivo delivery of saRNA.
Inside out: optimization of lipid nanoparticle formulations for exterior complexation and in vivo delivery of saRNA.
Gene therapy (2019-07-14)
Anna K Blakney, Paul F McKay, Bárbara Ibarzo Yus, Yoann Aldon, Robin J Shattock
ABSTRACT
Self-amplifying RNA (saRNA) is a promising biotherapeutic tool that has been used as a vaccine against both infectious diseases and cancer. saRNA has been shown to induce protein expression for up to 60 days and elicit immune responses with lower dosing than messenger RNA (mRNA). Because saRNA is a large (~9500 nt), negatively charged molecule, it requires a delivery vehicle for efficient cellular uptake and degradation protection. Lipid nanoparticles (LNPs) have been widely used for RNA formulations, where the prevailing paradigm is to encapsulate RNA within the particle, including the first FDA-approved small-interfering siRNA therapy. Here, we compared LNP formulations with cationic and ionizable lipids with saRNA either on the interior or exterior of the particle. We show that LNPs formulated with cationic lipids protect saRNA from RNAse degradation, even when it is adsorbed to the surface. Furthermore, cationic LNPs deliver saRNA equivalently to particles formulated with saRNA encapsulated in an ionizable lipid particle, both in vitro and in vivo. Finally, we show that cationic and ionizable LNP formulations induce equivalent antibodies against HIV-1 Env gp140 as a model antigen. These studies establish formulating saRNA on the surface of cationic LNPs as an alternative to the paradigm of encapsulating RNA.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
N,N-Dimethyldodecylamine, 97%