Merck
  • Application of D-Amino Acids as Biofilm Dispersing Agent in Dental Unit Waterlines
Application of D-Amino Acids as Biofilm Dispersing Agent in Dental Unit Waterlines
International journal of dentistry (2018)
Ampornaramveth RS, et al.