Merck
  • Using supported bilayers to study the spatiotemporal organization of membrane-bound proteins.
Using supported bilayers to study the spatiotemporal organization of membrane-bound proteins.
Methods in cell biology (2015-05-23)
Phuong A Nguyen, Christine M Field, Aaron C Groen, Timothy J Mitchison, Martin Loose
ABSTRACT
Cell division in prokaryotes and eukaryotes is commonly initiated by the well-controlled binding of proteins to the cytoplasmic side of the cell membrane. However, a precise characterization of the spatiotemporal dynamics of membrane-bound proteins is often difficult to achieve in vivo. Here, we present protocols for the use of supported lipid bilayers to rebuild the cytokinetic machineries of cells with greatly different dimensions: the bacterium Escherichia coli and eggs of the vertebrate Xenopus laevis. Combined with total internal reflection fluorescence microscopy, these experimental setups allow for precise quantitative analyses of membrane-bound proteins. The protocols described to obtain glass-supported membranes from bacterial and vertebrate lipids can be used as starting points for other reconstitution experiments. We believe that similar biochemical assays will be instrumental to study the biochemistry and biophysics underlying a variety of complex cellular tasks, such as signaling, vesicle trafficking, and cell motility.
Product Description
Avanti
Brain PI(4,5)P2, Avanti Research - A Croda Brand
Avanti
Brain PI(4,5)P2, Avanti Research - A Croda Brand
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Avanti
E. coli Extract Polar, Avanti Research - A Croda Brand
Avanti
Brain PC, Avanti Research - A Croda Brand 840053C
Avanti
Brain PC, Avanti Research - A Croda Brand 840053P, powder
Avanti
E. coli Extract Polar, Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
Liver PI, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Liver PI, Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand