Merck
  • Anti-mouse properdin TSR 5/6 monoclonal antibodies block complement alternative pathway-dependent pathogenesis.
Anti-mouse properdin TSR 5/6 monoclonal antibodies block complement alternative pathway-dependent pathogenesis.
Monoclonal antibodies in immunodiagnosis and immunotherapy (2015-02-28)
Paula Bertram, Antonina M Akk, Hui-fang Zhou, Lynne M Mitchell, Christine T N Pham, Dennis E Hourcade
ABSTRACT
The complement alternative pathway (AP) is a major contributor to a broad and growing spectrum of diseases that includes age-related macular degeneration, atypical hemolytic uremic syndrome, and preeclampsia. As a result, there is much interest in the therapeutic disruption of AP activity. Properdin, the only positive regulator of the AP, is a particularly promising AP target. Several issues need to be clarified before the potential for properdin-directed therapy can be realized. In this report we use a portion of the mouse properdin protein, expressed in a bacterial system, to raise rabbit polyclonal and hamster monoclonal antibodies that block properdin-dependent pathogenesis. These antibodies, when employed with AP-dependent mouse disease models, can help evaluate the feasibility of properdin-directed therapy.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, purified by gel-filtration chromatography
Pricing and availability is not currently available.