Merck
  • ZMIZ1 preferably enhances the transcriptional activity of androgen receptor with short polyglutamine tract.
ZMIZ1 preferably enhances the transcriptional activity of androgen receptor with short polyglutamine tract.
PloS one (2011-09-29)
Xiaomeng Li, Chunfang Zhu, William H Tu, Nanyang Yang, Hui Qin, Zijie Sun
ABSTRACT
The androgen receptor (AR) is a ligand-induced transcription factor and contains the polyglutamine (polyQ) tracts within its N-terminal transactivation domain. The length of polyQ tracts has been suggested to alter AR transcriptional activity in prostate cancer along with other endocrine and neurologic disorders. Here, we assessed the role of ZMIZ1, an AR co-activator, in regulating the activity of the AR with different lengths of polyQ tracts as ARQ9, ARQ24, and ARQ35 in prostate cancer cells. ZMIZ1, but not ZMIZ2 or ARA70, preferably augments ARQ9 induced androgen-dependent transcription on three different androgen-inducible promoter/reporter vectors. A strong protein-protein interaction between ZMIZ1 and ARQ9 proteins was shown by immunoprecipitation assays. In the presence of ZMIZ1, the N and C-terminal interaction of the ARQ9 was more pronounced than ARQ24 and ARQ35. Both Brg1 and BAF57, the components of SWI/SNF complexes, were shown to be involved in the enhancement of ZMIZ1 on AR activity. Using the chromatin immunoprecipitation assays (ChIP), we further demonstrated a strong recruitment of ZMIZ1 by ARQ9 on the promoter of the prostate specific antigen (PSA) gene. These results demonstrate a novel regulatory role of ZMIZ1 in modulating the polyQ tract length of AR in prostate cancer cells.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Androgen Receptor Antibody, from rabbit, purified by affinity chromatography