Merck
Target discovery and validation in pancreatic cancer.
Methods in molecular biology (Clifton, N.J.) (2006-12-19)
Robert M Beaty, Mads Gronborg, Jonathan R Pollack, Anirban Maitra
ABSTRACT
Pancreatic cancer is a lethal disease and rational strategies for early detection and targeted therapies are urgently required to alleviate the dismal prognosis of this neoplasm. The use of global RNA and protein expression-profiling technologies, such as DNA microarrays, serial analysis of gene expression, and mass spectrometric analysis of proteins, have led to identification of cellular targets with considerable potential for clinical application and patient care. These studies underscore the importance of pursuing large-scale profiling of human cancers not only for furthering our understanding of the pathogenesis of these malignancies but also for developing strategies to improve patient outcomes.
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%