- Europium-labeled activity-based probe through click chemistry: absolute serine protease quantification using (153)Eu isotope dilution ICP/MS.
Europium-labeled activity-based probe through click chemistry: absolute serine protease quantification using (153)Eu isotope dilution ICP/MS.
Angewandte Chemie (International ed. in English) (2012-02-22)
Xiaowen Yan, Yacui Luo, Zhubao Zhang, Zhaoxin Li, Qiang Luo, Limin Yang, Bo Zhang, Haifeng Chen, Peiming Bai, Qiuquan Wang
PMID22344943
ABSTRACT
Click and analyze: the titled probe was synthesized by conjugating a sulfonyl fluoride and azido unit using click chemistry to give SF-Eu, which can react specifically with serine (Ser) in the active site of serine protease (SP). Combination of the method with (153)Eu-isotope dilution ICP/MS enables absolute protein quantification of active SPs in biological samples using only one (153)Eu(NO(3))(3) isotopic standard.