Merck
  • Delayed myelination and neurodevelopment in male seizure-prone versus seizure-resistant rats.
Delayed myelination and neurodevelopment in male seizure-prone versus seizure-resistant rats.
Epilepsia (2018-01-30)
Pragati Sharma, Kim L Powell, Mary E Wlodek, Terence J O'Brien, Krista L Gilby
ABSTRACT
Aberrant myelination and developmental delay have been reported in epilepsy. However, it is unclear whether these are linked to intrinsic mechanisms that support a predisposition toward seizures and the development of epilepsy. Thus, we compared rates of myelination and neurodevelopment in male rats selectively bred for enhanced susceptibility to kindling epileptogenesis (FAST) with male rats bred for resistance (SLOW). Myelin-specific gene expression was compared in the brainstem, cerebellum, and cerebral hemisphere of FAST and SLOW rats on postnatal days (PNDs) 5, 11, 17, 23, and 90 to determine strain-specific myelination rates. Myelin protein levels were also compared at PNDs 5 and 23 in the brainstem. Relative rates of neurodevelopment were evaluated between PNDs 5 and 21 using physical growth landmarks and neuromotor tests including righting reflex, cliff avoidance, negative geotaxis, and locomotor activity. Myelin-specific mRNA expression was significantly down-regulated in FAST rats on PNDs 5 and 11 in all 3 brain structures, indicating relatively delayed myelination. Likewise, corresponding protein levels were significantly lower in FAST brainstem on PND 5. Developmental delay was evident in the FAST strain such that only 9% of FAST pups, compared to 81% of SLOW, had open eyes by PND 13, locomotor activity was significantly reduced between PNDs 12 and 16, and neuromotor task acquisition was delayed between PNDs 5 and 10. Relative delays in myelination and neurodevelopment co-occurred in the seizure-prone FAST strain in the absence of seizures. These findings suggest these symptoms are not seizure-induced and may be mechanistically linked to an underlying pathophysiology supporting a predisposition toward developing epilepsy.
Sigma-Aldrich
Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody, clone 8-18C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Myelin Proteolipid Protein Antibody, CT, clone PLPC1, clone PLPC1, Chemicon®, from mouse
Sigma-Aldrich
Anti-α-Tubulin Antibody, clone DM1A, clone DM1A, Upstate®, from mouse
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®