Merck
  • Exploring the In Vivo Role of the Mitochondrial Calcium Uniporter in Brown Fat Bioenergetics.
Exploring the In Vivo Role of the Mitochondrial Calcium Uniporter in Brown Fat Bioenergetics.
Cell reports (2019-05-03)
Daniel Flicker, Yasemin Sancak, Eran Mick, Olga Goldberger, Vamsi K Mootha
ABSTRACT
The mitochondrial calcium uniporter has been proposed to coordinate the organelle's energetics with calcium signaling. Uniporter current has previously been reported to be extremely high in brown adipose tissue (BAT), yet it remains unknown how the uniporter contributes to BAT physiology. Here, we report the generation and characterization of a mouse model lacking Mcu, the pore forming subunit of the uniporter, specifically in BAT (BAT-Mcu-KO). BAT-Mcu-KO mice lack uniporter-based calcium uptake in BAT mitochondria but exhibit unaffected cold tolerance, diet-induced obesity, and transcriptional response to cold in BAT. Unexpectedly, we found in wild-type animals that cold powerfully activates the ATF4-dependent integrated stress response (ISR) in BAT and upregulates circulating FGF21 and GDF15, raising the hypothesis that the ISR partly underlies the pleiotropic effects of BAT on systemic metabolism. Our study demonstrates that the uniporter is largely dispensable for BAT thermogenesis and demonstrates activation of the ISR in BAT in response to cold.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Anti-phospho-PDHE1-A type I (Ser293) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Harris Hematoxylin Solution, Modified
Sigma-Aldrich
Oil Red O, certified by the BSC
Sigma-Aldrich
Anti-LRPPRC antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
PhosphoDetect Anti-PDH-E1α (pSer³⁰⁰) Rabbit pAb, liquid, Calbiochem®