Merck
  • Methylation of p53 by Set7/9 mediates p53 acetylation and activity in vivo.
Methylation of p53 by Set7/9 mediates p53 acetylation and activity in vivo.
Molecular cell (2008-02-19)
Julia K Kurash, Hong Lei, Qiong Shen, Wendy L Marston, Brian W Granda, Hong Fan, Daniel Wall, En Li, François Gaudet
ABSTRACT
The protein methyltransferase Set7/9 was recently shown to regulate p53 activity in cancer cells. However, the impact of Set7/9 on p53 function in vivo is unclear. To explore these issues, we created a null allele of Set7/9 in mice. Cells from Set7/9 mutant mice fail to methylate p53 K369, are unable to induce p53 downstream targets upon DNA damage, and are predisposed to oncogenic transformation. Importantly, we find that methylation of p53 by Set7/9 is required for the binding of the acetyltransferase Tip60 to p53 and for the subsequent acetylation of p53. We provide the first genetic evidence demonstrating that lysine methylation of p53 by Set7/9 is important for p53 activation in vivo and suggest a mechanistic link between methylation and acetylation of p53 through Tip60.
Sigma-Aldrich
Anti-Histidine Tagged Antibody, clone 4D11, clone 4D11, Upstate®, from mouse
Sigma-Aldrich
Anti-Acetylated Lysine Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-p53 (Ab-6) (Pantropic) Mouse mAb (DO-1), liquid, clone DO-1, Calbiochem®
Sigma-Aldrich
Anti-p53 (Ab-1) (Pantropic) Mouse mAb (PAb421), liquid, clone PAb421, Calbiochem®