Merck
Layer 2/3 Pyramidal Neurons Control the Gain of Cortical Output.
Cell reports (2018-09-13)
Michael Quiquempoix, Sophie L Fayad, Katia Boutourlinsky, Nathalie Leresche, Régis C Lambert, Thomas Bessaih
ABSTRACT
Initial anatomical and physiological studies suggested that sensory information relayed from the periphery by the thalamus is serially processed in primary sensory cortical areas. It is thought to propagate from layer 4 (L4) up to L2/3 and down to L5, which constitutes the main output of the cortex. However, more recent experiments point toward the existence of a direct processing of thalamic input by L5 neurons. Therefore, the role of L2/3 neurons in the sensory processing operated by L5 neurons is now highly debated. Using cell type-specific and reversible optogenetic manipulations in the somatosensory cortex of both anesthetized and awake mice, we demonstrate that L2/3 pyramidal neurons play a major role in amplifying sensory-evoked responses in L5 neurons. The amplification effect scales with the velocity of the sensory stimulus, indicating that L2/3 pyramidal neurons implement gain control in deep-layer neurons.
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse