Merck
  • Realistic environmental exposure to microplastics does not induce biological effects in the Pacific oyster Crassostrea gigas.
Realistic environmental exposure to microplastics does not induce biological effects in the Pacific oyster Crassostrea gigas.
Marine pollution bulletin (2019-10-28)
Messika Revel, Amélie Châtel, Hanane Perrein-Ettajani, Mélanie Bruneau, Farida Akcha, Rossana Sussarellu, Julien Rouxel, Katherine Costil, Priscilla Decottignies, Bruno Cognie, Fabienne Lagarde, Catherine Mouneyrac
ABSTRACT
The aim of the present study was to evaluate the presence and potential toxic effects of plastic fragments (<400 μm) of polyethylene and polypropylene on the Pacific oyster Crassostrea gigas. Oysters were exposed to environmentally relevant concentrations (0, 0.008, 10, 100 μg of particles/L) during 10 days, followed by a depuration period of 10 days in clean seawater. Effects of microplastics were evaluated on the clearance rate of organisms, tissue alteration, antioxidant defense, immune alteration and DNA damage. Detection and quantification of microplastics in oyster's tissues (digestive gland, gills and other tissues) and biodeposits using infrared microscopy were also conducted. Microplastics were detected in oyster's biodeposits following exposure to all tested concentrations: 0.003, 0.006 and 0.05 particles/mg of biodeposits in oysters exposed to 0.008, 10 and 100 μg of particles/L, respectively. No significant modulation of biological markers was measured in organisms exposed to microplastics in environmentally relevant conditions.
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Acid phosphatase Assay Kit, 1 kit sufficient for 1,000 assays (multiwell plates), 1 kit sufficient for 100 assays (tubes)
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)