Merck
  • TMEM16F inhibition limits pain-associated behavior and improves motor function by promoting microglia M2 polarization in mice.
TMEM16F inhibition limits pain-associated behavior and improves motor function by promoting microglia M2 polarization in mice.
Biochemical and biophysical research communications (2019-08-15)
Jing Zhao, Quan-You Gao
ABSTRACT
Spinal cord injury (SCI) leads to sensorimotor deficits and autonomic changes. Macrophages and microglia could be polarized into the classically activated pro-inflammatory M1 phenotype or the alternatively activated anti-inflammatory M2 phenotype. Transmembrane protein with unknown function 16F (TMEM16F) exhibits functional diversity and may contribute to microglial function. However, the effects of TMEM16F on the modulation of macrophage/microglial polarization are still not fully understood. In the study, TMEM16F up-regulation was detected after SCI in mice, and TMEM16F protein was found in macrophages/microglia in injured spinal cord sections. Depletion of TMEM16F improved motor function in male mice with SCI. M1-type macrophages/microglia accumulated in lower numbers in the injured spinal cord of TMEM16F-knockout (KO) mice. M2 polarization inhibited by SCI was improved in mice with TMEM16F deficiency. TMEM16F deletion also attenuated microglial/macrophage pro-inflammatory response. Furthermore, significant down-regulation of A disintegrin and metalloprotease 17 (ADAM17) was observed in TMEM16F-KO mice. Importantly, TMEM16F-promoted M1 polarization and -inhibited M1 polarization were largely associated with the suppression of ADAM17. Overall, our findings provided new insights into the regulatory mechanisms of macrophage/microglial polarization, thereby possibly facilitating the development of new therapeutic strategies for SCI through the regulation of TMEM16F/ADAM17 signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting human ANO6