Merck
  • A heteromeric cis-prenyltransferase is responsible for the biosynthesis of glycosyl carrier lipids in Methanosarcina mazei.
A heteromeric cis-prenyltransferase is responsible for the biosynthesis of glycosyl carrier lipids in Methanosarcina mazei.
Biochemical and biophysical research communications (2019-10-09)
Koh-Ichi Emi, Kitty Sompiyachoke, Miyako Okada, Hisashi Hemmi
ABSTRACT
Cis-prenyltransferases are enzymes responsible for the biosynthesis of glycosyl carrier lipids, natural rubber, and some secondary metabolites. Certain organisms, including some archaeal species, possess multiple genes encoding cis-prenyltransferase homologs, and the physiological roles of these seemingly-redundant genes are often obscure. Cis-prenyltransferases usually form homomeric complexes, but recent reports have demonstrated that certain eukaryotic enzymes are heteromeric protein complexes consisting of two homologous subunits. In this study, three cis-prenyltransferase homolog proteins, MM_0014, MM_0618, and MM_1083, from the methanogenic archaeon Methanosarcina mazei are overexpressed in Escherichia coli and partially purified for functional characterization. Coexistence of MM_0618 and MM_1083 exhibits prenyltransferase activity, while each of them alone has almost no activity. The chain-lengths of the products of this heteromeric enzyme are in good agreement with those of glycosyl carrier lipids extracted from M. mazei, which are likely di- and tetra-hydrogenated decaprenyl phosphates, suggesting that the MM_0618/MM_1083 heteromer is involved in glycosyl carrier lipid biosynthesis. MM_0014 acts as a typical homomeric cis-prenyltransferase and produces shorter products.
Sigma-Aldrich
γ,γ-Dimethylallyl pyrophosphate triammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥90% (TLC)
Sigma-Aldrich
Isopentenyl pyrophosphate triammonium salt solution, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
Sigma-Aldrich
Geranylgeranyl pyrophosphate ammonium salt, solution, ≥95% (TLC), ~1 mg/mL in methanol: NH4OH (7:3)
Sigma-Aldrich
Farnesyl pyrophosphate ammonium salt, methanol:ammonia solution, ≥95% (TLC)