Merck
Energy extraction from nuts: walnuts, almonds and pistachios.
The British journal of nutrition (2019-10-18)
B M McArthur, R D Mattes
ABSTRACT
The bioaccessibility of fat has implications for satiety and postprandial lipidaemia. The prevailing view holds that the integrity of plant cell wall structure is the primary determinant of energy and nutrient extraction from plant cells as they pass through the gastrointestinal (GI) tract. However, comparisons across nuts (walnuts, almonds and pistachios) with varying physical properties do not support this view. In the present study, masticated samples of three nuts from healthy adults were exposed to a static model of gastric digestion followed by simulated intestinal digestion. Primary outcomes were particle size and lipid release at each phase of digestion. Walnuts produced a significantly larger particle size post-mastication compared with almonds. Under gastric and intestinal conditions, the particle size was larger for walnuts compared with pistachios and almonds (P < 0·05). However, the masticated and digesta particle sizes were not related to the integrity of cell walls or lipid release. The total lipid release was comparable between nuts after the in vitro intestinal phase (P > 0·05). Microstructural examination showed ruptured and fissured cell walls that would allow digestion of cellular contents, and this may be governed by internal cellular properties such as oil body state. Furthermore, the cell walls of walnuts tend to rupture rather than separate and as walnut tissue passes through the GI tract, lipids tend to coalesce reducing digestion efficiency.
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))