Merck
  • Safety Evaluation of PQ Birch Allergy Immunotherapy to Support Product Development.
Safety Evaluation of PQ Birch Allergy Immunotherapy to Support Product Development.
Regulatory toxicology and pharmacology : RTP (2019-08-20)
Paul Baldrick, James W Hutchings, Matthew D Heath, Murray A Skinner
ABSTRACT
PQ Birch represents an allergen-specific immunotherapy for the treatment of birch pollinosis. It consists of native birch pollen extract chemically modified with glutaldehyde adsorbed to L-tyrosine in its microcrystalline form with addition of the adjuvant Monophosphoryl Lipid A (MPL®). A nonclinical safety testing strategy was designed based upon interpretation of current legislation and regulatory intelligence and comprised genotoxicity studies (bacterial reverse mutation and Chinese hamster ovary micronucleus assays), a rat repeat dose toxicology study and a rabbit local tolerance study. No safety findings of concern were found. Thus, no evidence of genotoxicity was found. Relatively minor, immunostimulatory effects were seen following repeated subcutaneous dosing (once every 2 weeks for 13 weeks) as reversible increased white cell count (notably neutrophils), increased globulin level (resulting in decreased albumin/globulin [A/G] ratio) and increased fibrinogen, as well as minor dose site reaction in the form of inflammatory cell infiltrate. These findings are likely due to the immunostimulatory nature of MPL® and/or the presence of L-tyrosine within the adjuvanted vaccine. Similar dose site inflammatory changes to the injected formulation were also noted in the rabbit local tolerance study.
Sigma-Aldrich
2-Nitrofluorene, 98%
Sigma-Aldrich
N-(Cyanomethyl)acetamide, ≥97.0% (elemental analysis)
Supelco
9-Aminoacridine, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC)