Merck
  • Ubiquitin-specific peptidase 46 (USP46) suppresses renal cell carcinoma tumorigenesis through AKT pathway inactivation.
Ubiquitin-specific peptidase 46 (USP46) suppresses renal cell carcinoma tumorigenesis through AKT pathway inactivation.
Biochemical and biophysical research communications (2019-09-23)
Dingwen Gui, Wei Peng, Weidong Jiang, Geng Huang, Gang Liu, Zhihua Ye, Yang Wang, Zuwei Xu, Jinlun Fu, Shuai Luo, Yunfei Zhao
ABSTRACT
USP46, a member of the ubiquitin-specific protease family, plays essential roles in cancer cell proliferation and metastasis and is used as a candidate target for cancer therapeutics. However, the effects of USP46 on renal cell carcinoma (RCC) and its underlying molecular mechanism remain unknown. In this study, the predictive and prognostic relevance of USP46 in RCC, patient-derived primary tissues, and normal liver tissues obtained from the TCGA dataset were analyzed for the USP46 mRNA levels or prognostic relevance. Gain-of-function or loss-of-function assays were used to evaluate the vital roles of USP46 in tumor cell proliferation and cell migration. As a result, the USP46 expression level in RCC is highly decreased compared to normal tissues, and the Kaplan-Meier curve showed that USP46 high expression patients had good prognoses. Functionally, the forced expression of USP46 significantly restrained tumor cell proliferation, colony formation, and cell migration. The shRNA mediated USP46 knockdown cells exhibited the opposite results. We further showed that ectopically expressed USP46 obviously inhibited the AKT signaling pathway in cancer cells, while USP46 depletion caused a dramatic increase in AKT activity reflected by phosphorylation in the serine and threonine residues of AKT or downstream p70S6K1. Importantly, MK2206, a specific AKT inhibitor, completely counteracted the effects on cell proliferation, cell migration, and AKT activity in the USP46 depletion cells. We thus revealed a novel mechanism of USP46 regulation in RCC, and our data indicate that USP46 is a tumor suppressor in RCC via AKT signaling pathway inactivation.
Product Description
Polyetherimide, (PEI), film, thickness 0.175 mm, L 1 m, clear amber