Merck
  • Crystal structure of peptide-bound neprilysin reveals key binding interactions.
Crystal structure of peptide-bound neprilysin reveals key binding interactions.
FEBS letters (2019-09-13)
Stephen Moss, Vasanta Subramanian, K Ravi Acharya
ABSTRACT
Neprilysin (NEP) is a promiscuous zinc metalloprotease with broad substrate specificity and cleaves a remarkable diversity of substrates through endopeptidase action. Two of these - amyloid-β and natriuretic peptides - implicate the enzyme in both Alzheimer's disease and cardiovascular disease, respectively. Here, we report the creation of a catalytically inactive NEP (E584D) to determine the first peptide-bound crystal structure at 2.6 Å resolution. The structure reveals key interactions involved in substrate binding which we have identified to be conserved in other known zinc metalloproteases. In addition, the structure provides evidence for a potential exosite within the central cavity that may play a critical role in substrate positioning. Together, these results contribute to our understanding of the molecular function of NEP.
Sigma-Aldrich
Natriuretic Peptide, C-Type, ≥95% (HPLC)
Sigma-Aldrich
1-Ethyl-2-pyrrolidone, 98%
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile