Merck
  • Printed and flexible biosensor for antioxidants using interdigitated ink-jetted electrodes and gravure-deposited active layer
Printed and flexible biosensor for antioxidants using interdigitated ink-jetted electrodes and gravure-deposited active layer
Biosensors And Bioelectronics (2015)
Pavinatto FJ, et al.