Merck
  • Hydroxylation of guanine in nucleosides and DNA at the C-8 position by heated glucose and oxygen radical-forming agents.
Hydroxylation of guanine in nucleosides and DNA at the C-8 position by heated glucose and oxygen radical-forming agents.
Environmental health perspectives (1986-08-01)
H Kasai, S Nishimura
ABSTRACT
Heated glucose is mutagenic to Salmonella typhimurium TA 100 in the absence of S-9 mix. For identifying unknown mutagens in heated glucose (dry solid, 200 degrees C, 20 min), reaction with isopropylideneguanosine (IPG) was followed by isolation and characterization of the mutagen-IPG adduct. Two adducts, glyoxal-IPG and 8-hydroxy-IPG, were identified in the reaction mixture by this technique. To elucidate the mechanism of this hydroxylation reaction, we investigated the abilities of various agents to hydroxylate deoxyguanosine or guanine base in DNA. Various reducing agents, metals, asbestoses, polyphenols, aminophenols, and X-ray were effective for hydroxylation, and an oxygen radical seems to be the reactive species. For sensitive detection of 8-hydroxyguanine, a monoclonal antibody for it was prepared.
Sigma-Aldrich
2′,3′-O-Isopropylideneguanosine
Pricing and availability is not currently available.