Merck
  • Intracellular Ca2+ regulation of H+/Ca2+ antiporter YfkE mediated by a Ca2+ mini-sensor.
Intracellular Ca2+ regulation of H+/Ca2+ antiporter YfkE mediated by a Ca2+ mini-sensor.
Proceedings of the National Academy of Sciences of the United States of America (2020-04-29)
Shuo Lu, Zhenlong Li, Alemayehu A Gorfe, Lei Zheng
ABSTRACT
The H+/Ca2+ (calcium ion) antiporter (CAX) plays an important role in maintaining cellular Ca2+ homeostasis in bacteria, yeast, and plants by promoting Ca2+ efflux across the cell membranes. However, how CAX facilitates Ca2+ balance in response to dynamic cytosolic Ca2+ perturbations is unknown. Here, we identified a type of Ca2+ "mini-sensor" in YfkE, a bacterial CAX homolog from Bacillus subtilis. The mini-sensor is formed by six tandem carboxylate residues within the transmembrane (TM)5-6 loop on the intracellular membrane surface. Ca2+ binding to the mini-sensor triggers the transition of the transport mode of YfkE from a high-affinity to a low-affinity state. Molecular dynamics simulation and fluorescence resonance energy transfer analysis suggest that Ca2+ binding to the mini-sensor causes an adjacent segment, namely, the exchanger inhibitory peptide (XIP), to move toward the Ca2+ translocation pathway to interact with TM2a in an inward-open cavity. The specific interaction was demonstrated with a synthetic peptide of the XIP, which inhibits YfkE transport and interrupts conformational changes mediated by the mini-sensor. By comparing the apo and Ca2+-bound CAX structures, we propose the following Ca2+ transport regulatory mechanism of YfkE: Ca2+ binding to the mini-sensor induces allosteric conformational changes in the Ca2+ translocation pathway via the XIP, resulting in a rearrangement of the Ca2+-binding transport site in the midmembrane. Since the Ca2+ mini-sensor and XIP sequences are also identified in other CAX homologs and/or Ca2+ transporters, including the mammalian Na+/Ca2+ exchanger (NCX), our study provides a regulatory mechanism for the Ca2+/cation transporter superfamily.
Product Description
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand