Merck
HIV-1 Envelope and MPER Antibody Structures in Lipid Assemblies.
Cell reports (2020-04-30)
Kimmo Rantalainen, Zachary T Berndsen, Aleksandar Antanasijevic, Torben Schiffner, Xi Zhang, Wen-Hsin Lee, Jonathan L Torres, Lei Zhang, Adriana Irimia, Jeffrey Copps, Kenneth H Zhou, Young D Kwon, William H Law, Chaim A Schramm, Raffaello Verardi, Shelly J Krebs, Peter D Kwong, Nicole A Doria-Rose, Ian A Wilson, Michael B Zwick, John R Yates, William R Schief, Andrew B Ward
ABSTRACT
Structural and functional studies of HIV envelope glycoprotein (Env) as a transmembrane protein have long been complicated by challenges associated with inherent flexibility of the molecule and the membrane-embedded hydrophobic regions. Here, we present approaches for incorporating full-length, wild-type HIV-1 Env, as well as C-terminally truncated and stabilized versions, into lipid assemblies, providing a modular platform for Env structural studies by single particle electron microscopy. We reconstitute a full-length Env clone into a nanodisc, complex it with a membrane-proximal external region (MPER) targeting antibody 10E8, and structurally define the full quaternary epitope of 10E8 consisting of lipid, MPER, and ectodomain contacts. By aligning this and other Env-MPER antibody complex reconstructions with the lipid bilayer, we observe evidence of Env tilting as part of the neutralization mechanism for MPER-targeting antibodies. We also adapt the platform toward vaccine design purposes by introducing stabilizing mutations that allow purification of unliganded Env with a peptidisc scaffold.
Sigma-Aldrich
n-Dodecyl β-D-maltoside, ≥98% (GC)
Sigma-Aldrich
Cholesteryl hemisuccinate
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand