Merck
  • Synthesis of some N-galactosides of 3-aryl-5-benzyl (or substituted benzyl)-1,2,4-triazin-6(1H)-/ones or thiones of expected biological activity.
Synthesis of some N-galactosides of 3-aryl-5-benzyl (or substituted benzyl)-1,2,4-triazin-6(1H)-/ones or thiones of expected biological activity.
Nucleosides, nucleotides & nucleic acids (2003-10-10)
Abdel Kader Mansour, Mohga M Eid, Nasser S A M Khalil
ABSTRACT
The 1-(2,3,4,6-tetra-O-acetyl-beta-D-galactopyranosyl)-3-aryl-5-benzyl (or substituted benzyl)-1,2,4-triazin-6(1H)-/ones or thiones were prepared via galactosidation of 3-aryl-5-benzyl (or substituted benzyl)-1,2,4-triazin-6(1H)-/ones or thiones with 2,3,4,6-tetra-O-acetyl-alpha-D-galactopyranosyl bromide. The structure of the new galactosyl derivatives was based on both spectroscopic and chemical evidences.
Sigma-Aldrich
Acetobromo-α-D-galactose, ≥93% (TLC)
Pricing and availability is not currently available.