Merck
  • A novel optical probe based on d-penicillamine-functionalized graphene quantum dots: Preparation and application as signal amplification element to minoring of ions in human biofluid.
A novel optical probe based on d-penicillamine-functionalized graphene quantum dots: Preparation and application as signal amplification element to minoring of ions in human biofluid.
Journal of molecular recognition : JMR (2020-02-27)
Paria Pashazadeh-Panahi, Mohammad Hasanzadeh, Reza Eivazzadeh-Keihan
ABSTRACT
In this study, d-penicillamine-functionalized graphene quantum dots (DPA-GQD) has been synthesized, which significantly increases the fluorescence intensity of GQD. We used this simple fluorescent probe for metal ions detection in human plasma samples. Designed DPA-GQD respond to Hg2+ , Cu2+ , Au2+ , Ag+ , Co2+ , Zn2+ , and Pb2+ with high sensitivity. The fluorescence intensity of this probe decreased significantly in the presence of metal ions such as, Hg2+ , Cu2+ , Au2+ , Ag+ , Co2+ , Zn2+ , and Pb2+ . In this work, a promising probe for ions monitoring was introduced. Moreover, DPA-GQD probe has been tested in plasma samples. The functionalized DPA-GQDs exhibits great promise as an alternative to previous fluorescent probes for bio-labeling, sensing, and other biomedical applications in aqueous solution.
Sigma-Aldrich
2-(Diisopropylamino)ethyl methacrylate, 97%, contains ~100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
D-Penicillamine, 98-101%