Merck
Using proximity ligation assay to detect protein arginine methylation.
Methods (San Diego, Calif.) (2019-09-10)
Coralie Poulard, Julien Jacquemetton, Thuy Ha Pham, Muriel Le Romancer
ABSTRACT
Arginine methylation is now recognized as a major contributor to proteome diversity and is, as such, involved in a large range of cellular processes. There is a growing need for assessing endogenous protein arginine methylation in cells. Besides the classical immunoprecipitation, in situ proximity ligation assay (PLA) is a useful technique allowing at the same time the detection, localization and quantification of arginine methylation of a given protein within a cellular context. Here, we described in depth a standard PLA protocol applied to the detection of arginine methylation in combination with RNA interference and specific methyltransferase inhibitors. We demonstrated that the glucocorticoid receptor is methylated by the arginine methyltransferase PRMT5 inside the nucleus of MCF-7 cells. In addition, the automated quantification of protein arginine methylation performed using Image J is reported. Hence, we demonstrated that PLA offers a novel approach to study protein arginine methylation and could be extended to other post-translational modifications when specific antibodies are available.
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)