Merck
  • Genetic and genomic characterization of vulva size traits in Yorkshire and Landrace gilts.
Genetic and genomic characterization of vulva size traits in Yorkshire and Landrace gilts.
BMC genetics (2020-03-14)
Flor-Anita Corredor, Leticia P Sanglard, Richard J Leach, Jason W Ross, Aileen F Keating, Nick V L Serão
ABSTRACT
Reproductive performance is critical for efficient swine production. Recent results indicated that vulva size (VS) may be predictive of reproductive performance in sows. Study objectives were to estimate genetic parameters, identify genomic regions associated, and estimate genomic prediction accuracies (GPA) for VS traits. Heritability estimates of VS traits, vulva area (VA), height (VH), and width (VW) measurements, were moderately to highly heritable in Yorkshire, with 0.46 ± 0.10, 0.55 ± 0.10, 0.31 ± 0.09, respectively, whereas these estimates were low to moderate in Landrace, with 0.16 ± 0.09, 0.24 ± 0.11, and 0.08 ± 0.06, respectively. Genetic correlations within VS traits were very high for both breeds, with the lowest of 0.67 ± 0.29 for VH and VW for Landrace. Genome-wide association studies (GWAS) for Landrace, reveled genomic region associated with VS traits on Sus scrofa chromosome (SSC) 2 (154-157 Mb), 7 (107-110 Mb), 8 (4-6 Mb), and 10 (8-19 Mb). For Yorkshire, genomic regions on SSC 1 (87-91 and 282-287 Mb) and 5 (67 Mb) were identified. All regions explained at least 3.4% of the genetic variance. Accuracies of genomic prediction were moderate in Landrace, ranging from 0.30 (VH) to 0.61 (VA), and lower for Yorkshire, with 0.07 (VW) to 0.11 (VH). Between-breed and multi-breed genomic prediction accuracies were low. Our findings suggest that VS traits are heritable in Landrace and Yorkshire gilts. Genomic analyses show that major QTL control these traits, and they differ between breed. Genomic information can be used to increase genetic gains for these traits in gilts. Additional research must be done to validate the GWAS and genomic prediction results reported in our study.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %