- Characterization of a recombinant L-fucose isomerase from Caldicellulosiruptor saccharolyticus that isomerizes L-fucose, D-arabinose, D-altrose, and L-galactose.
Characterization of a recombinant L-fucose isomerase from Caldicellulosiruptor saccharolyticus that isomerizes L-fucose, D-arabinose, D-altrose, and L-galactose.
Biotechnology letters (2009-10-27)
Yo-Han Ju, Deok-Kun Oh
PMID19856146
ABSTRACT
A recombinant L-fucose isomerase from Caldicellulosiruptor saccharolyticus was purified as a single 68 kDa band with an activity of 76 U mg(-1). The molecular mass of the native enzyme was 204 kDa as a trimer. The maximum activity for L-fucose isomerization was at pH 7 and 75 degrees C in the presence of 1 mM Mn(2+). Its half-life at 70 degrees C was 6.1 h. For aldose substrates, the enzyme displayed activity in decreasing order for L-fucose, with a k (cat) of 11,910 min(-1) and a K (m) of 140 mM, D-arabinose, D-altrose, and L-galactose. These aldoses were converted to the ketoses L-fuculose, D-ribulose, D-psicose, and L-tagatose, respectively, with 24, 24, 85, 55% conversion yields after 3 h.