Merck
  • The immunosuppressive functions of two novel tick serpins, HlSerpin-a and HlSerpin-b, from Haemaphysalis longicornis.
The immunosuppressive functions of two novel tick serpins, HlSerpin-a and HlSerpin-b, from Haemaphysalis longicornis.
Immunology (2019-10-14)
Fanqi Wang, Zhenyu Song, Jing Chen, Qihan Wu, Xia Zhou, Xiaohua Ni, Jianfeng Dai
ABSTRACT
Serpins are evolutionarily conserved serine protease inhibitors that are widely distributed in animals, plants and microbes. In this study, we reported the cloning and functional characterizations of two novel serpin genes, HlSerpin-a and HlSerpin-b, from the hard tick Haemaphysalis longicornis of China. Recombinant HlSerpin-a and HlSerpin-b displayed protease inhibitory activities against multiple mammalian proteases. Similar to other tick serpins, HlSerpin-a and HlSerpin-b suppressed the expression of inflammatory cytokines such as TNF-α, interleukin (IL)-6 and IL-1β from lipopolysaccharide-stimulated mouse bone-marrow-derived macrophages (BMDMs) or mouse bone-marrow-derived dendritic cells (BMDCs). The minimum active region (reaction centre loop) of HlSerpin-a, named SA-RCL, showed similar biological activities as HlSerpin-a in the protease inhibition and immune suppression assays. The immunosuppressive activities of full-length HlSerpin-a and SA-RCL are impaired in Cathepsin G or Cathepsin B knockout mouse macrophages, suggesting that the immunomodulation functions of SA and SA-RCL are dependent on their protease inhibitory activity. Finally, we showed that both full-length HlSerpins and SA-RCL can relieve the joint swelling and inflammatory response in collagen-induced mouse arthritis models. These results suggested that HlSerpin-a and HlSerpin-b are two functional arthropod serpins, and the minimal reactive peptide SA-RCL is a potential candidate for drug development against inflammatory diseases.