Merck
  • Discerning the inefficacy of hydroxyl radicals during perfluorooctanoic acid degradation.
Discerning the inefficacy of hydroxyl radicals during perfluorooctanoic acid degradation.
Chemosphere (2020-01-25)
Hassan Javed, Cong Lyu, Ruonan Sun, Danning Zhang, Pedro J J Alvarez
ABSTRACT
Perfluorooctanoic acid (PFOA) is a recalcitrant contaminant of emerging concern, and there is growing interest in advanced oxidation processes to degrade it. However, there is ambiguity in the literature about the efficacy of hydroxyl radicals (OH) for degrading PFOA. Here, we resolve this controversy by comparing PFOA degradation by UV photolysis (254 nm, 6 × 10-6 E/L.s) versus UV + H2O2, which produces OH. We optimized OH production in a UV + H2O2 system using nitrobenzene (NB) as a OH probe, but even under optimized conditions (i.e., 5 g/L H2O2), no significant difference occurred in PFOA removal by UV photolysis (21.1 ± 0.4%) versus UV + H2O2 (19.7 ± 0.7%) after 1-day treatment. Both treatments also resulted in similar daughter by-product concentrations and defluorination efficiencies (9.5 ± 1.7% for UV photolysis and 6.8 ± 1.0% for UV + H2O2), which indicates that OH is ineffective towards PFOA degradation and infers that other degradation mechanisms that are independent of OH production should be explored.
Sigma-Aldrich
Perfluoroheptanoic acid, ≥97.0%
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Undecafluorohexanoic acid, ≥97.0% (T)
Sigma-Aldrich
Pentafluoropropionic acid, 97%
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Sigma-Aldrich
Perfluoropentanoic acid, 97%