Merck
  • Development and optimization of lyophilized orally disintegrating tablets using factorial design.
Development and optimization of lyophilized orally disintegrating tablets using factorial design.
Pharmaceutical Development and Technology (2011)
Ahmed IS, Shamma RN, Shoukri RA.