Merck
  • Docosahexaenoic acid protects motor function and increases dopamine synthesis in a rat model of Parkinson's disease via mechanisms associated with increased protein kinase activity in the striatum.
Docosahexaenoic acid protects motor function and increases dopamine synthesis in a rat model of Parkinson's disease via mechanisms associated with increased protein kinase activity in the striatum.
Neuropharmacology (2020-02-01)
Neha Milind Chitre, Bo Jarrett Wood, Azizi Ray, Nader H Moniri, Kevin Sean Murnane
ABSTRACT
Parkinson's disease (PD) is a devastating neurodegenerative disease that leads to motor deficits and selective destruction of nigrostriatal dopaminergic neurons. PD is typically treated by dopamine replacement agents; however, dopamine replacement loses effectiveness in the later stages of the disease. Here, we describe the neuroprotective effects of the omega-3 fatty acid docosahexaenoic acid (DHA) in the medial forebrain bundle 6-hydroxydopamine (6-OHDA) model of advanced-stage PD in rats. We show that daily administration of DHA protects against core symptoms of PD, including deficits in postural stability, gait integrity, and dopamine neurochemistry in motor areas of the striatum. Our results also demonstrate that DHA increases striatal dopamine synthesis via phosphorylation of the rate-limiting catecholamine synthesizing enzyme tyrosine hydroxylase, in a manner dependent on the second messenger-linked protein kinases PKA and PKC. We also show that DHA specifically reverses dopamine loss in the nigrostriatal pathway, with no effect in the mesolimbic or mesocortical pathways. This suggests that DHA is unlikely to produce pharmacotherapeutic or adverse effects that depend on dopamine pathways other than the nigrostriatal pathway. To our knowledge, previous reports have not examined the effects of DHA in such an advanced-stage model, documented that the dopamine synthesizing effects of DHA in vivo are mediated through the activation of protein kinases and regulation of TH activity, or demonstrated specificity to the nigrostriatal pathway. These novel findings corroborate the beneficial effects of omega-3 fatty acids seen in PD patients and suggest that DHA provides a novel means of protecting patients for dopamine neurodegeneration.
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
H-89 dihydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Bisindolylmaleimide II, ≥97% (Mixture of 2 isomers)
Sigma-Aldrich
Desipramine hydrochloride, ≥98% (TLC), powder
Sigma-Aldrich
6-Hydroxydopamine hydrochloride, ≥97% (titration), powder