Merck
  • Inkjet-printed electrochemically reduced graphene oxide microelectrode as a platform for HT-2 mycotoxin immunoenzymatic biosensing.
Inkjet-printed electrochemically reduced graphene oxide microelectrode as a platform for HT-2 mycotoxin immunoenzymatic biosensing.
Biosensors & bioelectronics (2020-04-11)
Jiri Kudr, Lei Zhao, Emily P Nguyen, Henri Arola, Tarja K Nevanen, Vojtech Adam, Ondrej Zitka, Arben Merkoçi
ABSTRACT
The design and application of an inkjet-printed electrochemically reduced graphene oxide microelectrode for HT-2 mycotoxin immunoenzymatic biosensing is reported. A water-based graphene oxide ink was first formulated and single-drop line working microelectrodes were inkjet-printed onto poly(ethylene 2,6-naphthalate) substrates, with dimensions of 78 μm in width and 30 nm in height after solvent evaporation. The printed graphene oxide microelectrodes were electrochemically reduced and characterized by Raman and X-ray photoelectron spectroscopies in addition to microscopies. Through optimization of the electrochemical reduction parameters, differential pulse voltammetry were performed to examine the sensing of 1-naphthol (1-N), where it was revealed that reduction times had significant effects on electrode performance. The developed microelectrodes were then used as an immunoenzymatic biosensor for the detection of HT-2 mycotoxin based on carbodiimide linking of the microelectrode surface and HT-2 toxin antigen binding fragment of antibody (anti-HT2 (10) Fab). The HT-2 toxin and anti-HT2 (10) Fab reaction was reported by anti-HT2 immune complex single-chain variable fragment of antibody fused with alkaline phosphatase (anti-IC-HT2 scFv-ALP) which is able to produce an electroactive reporter - 1-N. The biosensor showed detection limit of 1.6 ng ∙ mL-1 and a linear dynamic range of 6.3 - 100.0 ng ∙ mL-1 within a 5 min incubation with 1-naphthyl phosphate (1-NP) substrate.
Sigma-Aldrich
1-Naphthyl phosphate, 99%
Pricing and availability is not currently available.